Order Entry
Denmark
ContactUsLinkComponent
Anti-PDZD2 Goat Polyclonal Antibody
Anti-PDZD2 Goat Polyclonal Antibody
Catalog # PRSI46-129
Supplier:  ProSci Inc.
undefined
Anti-PDZD2 Goat Polyclonal Antibody
Catalog # PRSI46-129
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    PDZ Domain Containing 2
  • Antigen symbol:
    PDZD2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    PAPIN|PDZK3|PIN1|PDZ domain containing 3|PDZ domain-containing protein 3|PDZ domain-containing protein 2|AIPC|activated in prostate cancer protein
  • Concentration:
    500 ug/mL
  • Immunogen:
    PAPIN antibody was raised against a 14 amino acid synthetic peptide near the N-Terminus of PAPIN.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

PAPIN antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PAPIN antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PDZD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0