Anti-TRIM63 Goat Polyclonal Antibody
Catalog # PRSI45-918
Supplier: ProSci Inc.
Specifications
- Antigen name:Tripartite Motif Containing 63
- Antigen symbol:TRIM63
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:tripartite motif-containing 63|muscle specific ring finger protein 2|MURF1|IRF|tripartite motif-containing protein 63|SMRZ|RNF28|iris ring finger protein|striated muscle RING zinc finger protein|MURF2|E3 ubiquitin-protein ligase TRIM63|muscle-specific RING finger protein 1|ring finger protein 28
- Concentration:500 ug/mL
- Immunogen:MURF1 antibody was raised against a 15 amino acid synthetic peptide near the internal region of MURF1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
MURF1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
MURF1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRIM63
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human