Anti-NR0B1 Goat Polyclonal Antibody
PRSI45-461
- Antigen name:Nuclear Receptor Subfamily 0, Group B, Member 1
- Antigen symbol:NR0B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:AHC|AHCH|DAX1|NROB1|SRXY2|HHG|GTD|DAX-1|AHX|DSS
- Concentration:500 ug/mL
- Immunogen:DAX1 antibody was raised against a 13 amino acid synthetic peptide near the internal region of DAX1.
- Size:100 µg
- Pk:100 µG
DAX1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
DAX1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NR0B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human