Anti-LY6E Goat Polyclonal Antibody
PRSI42-628
- Antigen name:Lymphocyte Antigen 6 Complex, Locus E
- Antigen symbol:LY6E
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:RIGE|SCA-2|RIG-E|SCA2|TSA-1
- Concentration:500 ug/mL
- Immunogen:LY6E antibody was raised against an 11 amino acid synthetic peptide near the internal region of LY6E (mouse).
- Size:100 µg
- Pk:100 µG
LY6E antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
LY6E antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LY6E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: