Anti-PCSK9 Goat Polyclonal Antibody
Catalog # PRSI42-479
Supplier: ProSci Inc.
Specifications
- Antigen name:Proprotein convertase subtilisin/kexin type 9
- Antigen symbol:PCSK9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:PCSK9|Neural apoptosis-regulated convertase 1|NARC-1|NARC1|Proprotein convertase subtilisin/kexin type 9|Subtilisin/kexin-like protease PC9|Proprotein convertase PC9|PSEC0052
- Concentration:500 ug/mL
- Immunogen:PCSK9 antibody was raised against a 15 amino acid synthetic peptide near the internal region of PCSK9 (aa 214-228).
- Size:100 µg
- Pk:100 µG
Specifications
About this item
PCSK9 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
PCSK9 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PCSK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human