Anti-NAT6 Rabbit Polyclonal Antibody
PRSI30-441
- Antigen name:N-Acetyltransferase 6 (Gcn5-Related)
- Antigen symbol:NAT6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:FUS-2|FUS2
- Immunogen:A synthetic peptide corresponding a region of human NAT6.
- Size:50 µg
- Pk:50 µG
NAT6 belongs to the acetyltransferase family. It contains 1 N-acetyltransferase domain. NAT6 seems to be involved in N-acetylation. Acts on peptides with a N-terminal Met followed by Asp/Glu/Asn. It may act as a tumor suppressor. Defects in NAT6 are found in non-small cell lung cancer (NSCLC) cell lines.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NAT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human