Anti-ARIH1 Rabbit Polyclonal Antibody
Catalog # PRSI29-830
Supplier: ProSci Inc.
Specifications
- Antigen name:Ariadne RBR E3 Ubiquitin Protein Ligase 1
- Antigen symbol:ARIH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Zebrafish,Human,Dog,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:Drosophila|ariadne homolog|FLJ93118|ariadne ubiquitin-conjugating enzyme E2 binding protein homolog 1|1 (Drosophila)|FLJ20329|HHARI|ARIH1|ubiquitin-conjugating enzyme E2 binding protein|UBCH7BP|DKFZp686O13120|HARI|homolog of|ARI|ariadne
- Immunogen:A synthetic peptide corresponding a region of human ARIH1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ARIH1 might act as an E3 ubiquitin-protein ligase, or as part of the E3 complex, which accepts ubiquitin from specific E2 ubiquitin-conjugating enzymes, such as UBE2L3/UBCM4, and then transfers it to substrates.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ARIH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog, Zebrafish