Anti-AGER Rabbit Polyclonal Antibody
Product Details & Documents
AGER mediates interactions of advanced glycosylation end products (AGE). These are nonenzymatically glycosylated proteins which accumulate in vascular tissue in aging and at an accelerated rate in diabetes. It is receptor for amyloid beta peptide.This gene encodes a member of the immunoglobulin superfamily of cell surface molecules. It is a receptor for various molecules, including the amyloidogenic form of serum amyloid A, amyloid-beta protein, members of the S100/calgranulin superfamily and advanced glycation end products. The gene lies within the major histocompatibility complex (MHC) class III region on chromosome 6. Alternative splicing results in two transcript variants encoding different isoforms.This gene encodes a member of the immunoglobulin superfamily of cell surface molecules. It is a receptor for various molecules, including the amyloidogenic form of serum amyloid A, amyloid-beta protein, members of the S100/calgranulin superfamily and advanced glycation end products. The gene lies within the major histocompatibility complex (MHC) class III region on chromosome 6. Alternative splicing results in two transcript variants encoding different isoforms.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: AGER
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
10108-296.pdf
Datasheet
Specifikationer
- Size:50 µg
- Western blot:Yes
- Clonality:Polyclonal
- Form:Lyophilized
- Host:Rabbit
- Antigen name:Advanced Glycosylation End Product-specific Receptor
- Cat. No.:PRSI29-610
- Reactivity:Human
- Antigen synonyms:AGER
- Immunogen:A synthetic peptide corresponding a region of human AGER.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:AGER
- ELISA:Yes



