Anti-ZBTB32 Rabbit Polyclonal Antibody
PRSI27-428
- Antigen name:Zinc Finger And BTB Domain Containing 32
- Antigen symbol:ZBTB32
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Dog
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:FAXF|Rog|ZNF538|FAZF|TZFP
- Immunogen:A synthetic peptide corresponding a region of human ZBTB32.
- Size:50 µg
- Pk:50 µG
ZBTB32 may play an essential role during the proliferative stages of primitive hematopoietic progenitors, possibly acting in concert with (a subset of) the Fanconi anemia proteins. This gene can also interact with GATA-2 and can modify GATA-2 transactivation capacity.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZBTB32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human