- Antigen name:Syntaxin Binding Protein 1
- Antigen symbol:STXBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:FLJ37475|MUNC18-1|syntaxin-binding protein 1|rbSec1|syntaxin binding protein 1|RBSEC1|hUNC18|Hunc18|UNC18|RP11-56D16.3|EIEE4
- Immunogen:A synthetic peptide corresponding a region of human STXBP1.
- Size:50 µg
- Pk:50 µG
STXBP1 belongs to the STXBP/unc-18/SEC1 family. It may participate in the regulation of synaptic vesicle docking and fusion, possibly through interaction with GTP-binding proteins. The protein is essential for neurotransmission and binds syntaxin, a component of the synaptic vesicle fusion machinery probably in a 1:1 ratio. It can interact with syntaxins 1, 2, and 3 but not syntaxin 4.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: STXBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat