Anti-FUBP3 Rabbit Polyclonal Antibody
Catalog # PRSI26-610
Supplier: ProSci Inc.
Specifications
- Antigen name:Far upstream element-binding protein 3
- Antigen symbol:FUBP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:Far upstream element-binding protein 3|FUBP3|FUSE-binding protein 3
- Immunogen:A synthetic peptide corresponding a region of human FUBP3.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
FUBP3 may interact with single-stranded DNA from the far-upstream element (FUSE). FUBP3 may activate gene expression.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: FUBP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human