Anti-PTPLAD1 Rabbit Polyclonal Antibody
PRSI26-199
- Antigen name:Protein Tyrosine Phosphatase-like A Domain Containing 1
- Antigen symbol:PTPLAD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human PTPLAD1.
- Size:50 µg
- Pk:50 µG
PTPLAD1 is a multi-pass membrane protein. It belongs to the PTPLA family and contains 1 CS domain. PTPLAD1 (hB-ind1) plays a crucial role in HCV RNA replication and the propagation of JFH1 virus through interaction with viral and host proteins. It is involved in Rac1-signaling pathways leading to the modulation of gene expression.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PTPLAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat