Specifications
- Antigen name:Solute Carrier Family 6 (neutral amino acid transporter) Member 15
- Antigen symbol:SLC6A15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:NTT73|Transporter v7-3|Sodium-coupled branched-chain amino-acid transporter 1|SLC6A15|SBAT1|Solute carrier family 6 member 15|Sodium-dependent neutral amino acid transporter B(0)AT2|B0AT2
- Immunogen:A synthetic peptide corresponding a region of human SLC6A15.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SLC6A15 shows structural characteristics of an Na (+) and Cl (-)-dependent neurotransmitter transporter, including 12 transmembrane (TM) domains, intracellular N and C termini, and large extracellular loops containing multiple N-glycosylation sites.SLC6A15 shows structural characteristics of an Na (+) and Cl (-)-dependent neurotransmitter transporter, including 12 transmembrane (TM) domains, intracellular N and C termini, and large extracellular loops containing multiple N-glycosylation sites (Farmer et al., 2000 [PubMed 11112352]).
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC6A15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human