- Antigen name:Solute Carrier Family 39 Member A7
- Antigen symbol:SLC39A7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Dog
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:HKE4|H2-KE4|D6S115E|Zinc transporter SLC39A7|D6S2244E|RING5|Histidine-rich membrane protein Ke4|KE4|ZIP7
- Immunogen:A synthetic peptide corresponding a region of human SLC39A7.
- Size:50 µg
- Pk:50 µG
Zinc is an essential cofactor for more than 50 classes of enzymes. It is involved in protein, nucleic acid, carbohydrate, and lipid metabolism, as well as in the control of gene transcription, growth, development, and differentiation. Zinc cannot passively diffuse across cell membranes and requires specific transporters, such as SLC39A7, to enter the cytosol from both the extracellular environment and from intracellular storage compartments.Zinc is an essential cofactor for more than 50 classes of enzymes. It is involved in protein, nucleic acid, carbohydrate, and lipid metabolism, as well as in the control of gene transcription, growth, development, and differentiation. Zinc cannot passively diffuse across cell membranes and requires specific transporters, such as SLC39A7, to enter the cytosol from both the extracellular environment and from intracellular storage compartments.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC39A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human