- Antigen name:Arginine-Glutamic Acid Dipeptide (Re) Repeats
- Antigen symbol:RERE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:ARG|DNB1|ARP|ATN1L
- Immunogen:A synthetic peptide corresponding a region of human RERE.
- Size:50 µg
- Pk:50 µG
RERE is a member of the atrophin family of arginine-glutamic acid (RE) dipeptide repeat-containing proteins. RERE co-localizes with a transcription factor in the nucleus, and its overexpression triggers apoptosis. A similar protein in mouse associates with histone deacetylase and is thought to function as a transcriptional co-repressor during embryonic development.This gene encodes a member of the atrophin family of arginine-glutamic acid (RE) dipeptide repeat-containing proteins. The encoded protein co-localizes with a transcription factor in the nucleus, and its overexpression triggers apoptosis. A similar protein in mouse associates with histone deacetylase and is thought to function as a transcriptional co-repressor during embryonic development. Multiple transcript variants encoding different isoforms have been found for this gene.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RERE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat