Specifications
- Antigen name:Transcription Factor 7-like 2
- Antigen symbol:TCF7L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:TCF7L2|TCF4|Transcription factor 7-like 2|T-cell factor 4|TCF-4|T-cell-specific transcription factor 4|HMG box transcription factor 4|hTCF-4
- Immunogen:A synthetic peptide corresponding a region of human TCF7L2.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
The TCL7L2 is a high mobility group (HMG) box-containing transcription factor implicated in blood glucose homeostasis. The study of Yi et al. suggested that TCL7L2 acts through regulation of proglucagon through repression of the proglucagon gene in enteroendocrine cells via the Wnt signaling pathway. The TCL7L2 gene product is a high mobility group (HMG) box-containing transcription factor implicated in blood glucose homeostasis. The study of Yi et al. (2005) [PubMed 15525634] suggested that TCL7L2 acts through regulation of proglucagon (MIM 138030) through repression of the proglucagon gene in enteroendocrine cells via the Wnt signaling pathway.
TCF7L2 antibody can be used for detection of TCF7L2 by ELISA at 1:2500. TCF7L2 antibody can be used for detection of TCF7L2 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TCF7L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human