Order Entry
Denmark
ContactUsLinkComponent
Anti-MAFG Rabbit Polyclonal Antibody
Anti-MAFG Rabbit Polyclonal Antibody
Catalog # PRSI25-085
Supplier:  ProSci Inc.
undefined
Anti-MAFG Rabbit Polyclonal Antibody
Catalog # PRSI25-085
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    V-maf Musculoaponeurotic Fibrosarcoma Oncogene Homolog G
  • Antigen symbol:
    MAFG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Lyophilized
  • Immunogen:
    A synthetic peptide corresponding a region of human MAFG.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Since they lack a putative transactivation domain, the small Mafs behave as transcriptional repressors when they dimerize among themselves. However, they seem to serve as transcriptional activators by dimerizing with other (usually larger) basic-zipper proteins and recruiting them to specific DNA-binding sites. Small Maf proteins heterodimerize with Fos and may act as competitive repressors of the NF-E2 transcription factor. Transcription factor, component of erythroid-specific transcription factor NF-E2.

MAFG antibody can be used for detection of MAFG by ELISA at 1:62500. MAFG antibody can be used for detection of MAFG by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAFG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat