Anti-DCP2 Rabbit Polyclonal Antibody
Catalog # PRSI7049
Supplier: ProSci Inc.
Specifications
- Antigen name:Decapping mRNA 2
- Antigen symbol:DCP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:Nudix motif 20|Nucleoside diphosphate-linked moiety X motif 20|NUDT20|m7GpppN-mRNA hydrolase
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:DCP2 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human DCP2.
- Tested applications:E, WB, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
DCP2 Antibody: The removal, or decapping, of eukaryotic mRNA is an important step in the degradation of mRNA. Decapping protein 2 (DCP2) is the major mRNA decapping enzyme in cells. It is a member of the Nudix hydrolases superfamily of proteins that predominantly catalyze the hydrolysis of small nucleoside diphosphate substrates linked to another moiety. DCP2 is widely expressed in multiple tissues at varying levels, with highest expression seen in testis and brain.
Type:
Antigen: DCP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat