- Antigen name:WD Repeat Domain 18
- Antigen symbol:WDR18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:WDR18|Ipi3|R32184_1|WD repeat-containing protein 18
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:WDR18 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human WDR18.
- Tested applications:E, WB, IHC
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
WDR18 Antibody: WD repeat domain 18 (WDR18) is a member of the WD repeat protein family, which is involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. WD repeats are minimally conserved regions of approximately 40 amino acids that commonly form a tertiary propeller structure. WDR18 is a 432 amino acid protein that contains six WD repeats and is encoded by a gene located on human chromosome 19. WDR18 may play a role during development. The complex PELP1-TEX10-WDR18 is required for the maturation and nucleolar release of the large ribosomal subunit.
Type:
Antigen: WDR18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat