- Antigen name:Discs, Large (Drosophila) Homolog-Associated Protein 1
- Antigen symbol:DLGAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:SAP90/PSD-95-associated protein 1|beta isoform|DAP-1-ALPHA|PSD 95/SAP90 binding protein 1|PSD-95/SAP90-binding protein 1|DAP1|GKAP|DAP 1|Guanylate kinase-associated protein|GKPA/SAPAP|Dlgap1|Guanylate kinase associated protein|DAP-1-BETA|mKIAA4162|PSD95 binding protein 1|DAP-1|hGKAP|Disks large-associated protein 1|MGC88156|Disks large associated protein 1|included|DLGAP 1|KIAA4162|SAPAP1.|alpha isoform|GKAP/SAPAP|DAP 1 ALPHA|DAP 1 BETA|SAP90/PSD 95 associated protein 1|Discs large Drosophila homolog associated protein 1|DLGP1_HUMAN|SAPAP 1|Discs large homolog associated protein 1
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:SAPAP1 antibody was raised against a 18 amino acid synthetic peptide from near the center of human SAPAP1.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
SAPAP1 Antibody: SAP90/PSD-95-associated protein 1 (SAPAP1, also known as DLGAP1 or GKAP) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP1 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). SAPAP1 mRNA is targeted to cell bodies in a similar manner to SAPAP2 and -4, whereas SAPAP3 mRNA is detected mainly in cell bodies. SAPAP1 protein however, is targeted to the synapse and is not reliant on the synaptic localization of PSD-95 or the synaptic scaffolding molecule (S-SCAM).
Type:
Antigen: DLGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat