Specifications
- Antigen name:Nucleolar protein of 30 kDa
- Antigen symbol:NOP30
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:Nop 30|Nop30|Apoptosis repressor with CARD|Myp|Muscle-enriched cytoplasmic protein|NOL3|Nucleolar protein of 30 kDa|Nucleolar protein 3
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:Nop30 antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human Nop30.
- Tested applications:E, WB
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Nop30 Antibody: Apoptosis, also known as programmed cell death, plays major roles in development and normal tissue turnover in addition to tumor formation. Apoptosis is regulated by death domain (DD) and/or caspase recruitment domain (CARD) containing molecules and the caspase family of proteases. CARD domain containing cell death regulators include RAIDD, Apaf-1, caspase-9, and caspase-2. A novel CARD domain containing protein was recently identified and designated ARC for apoptosis repressor with CARD. An alternate splicing isoform of ARC was identified as Nop30. While ARC interacts with caspase-2 and -8 and suppresses apoptosis induced by cell death adapters FADD and TRADD and by cell death receptors Fas, TNFR-1 and DR3, Nop30 multimerizes and binds to the splicing factor SRp30c and may act to influence alternative splice site selection in vivo. The Nop30 antibody will not detect ARC protein.
Type:
Antigen: NOP30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat