Specifications
- Antigen name:TRAF3 interacting Protein 2
- Antigen symbol:CIKS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:CIKS antibody was raised against a synthetic peptide corresponding to amino acids 554 to 568 of human CIKS.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
CIKS Antibody: Nuclear factor kappa B (NF-kappa B) is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kappa B mediates the expression of a great variety of genes in response to extracellular stimuli. NF-kappa B associates with I kappa B proteins in the cell cytoplasm, which inhibit NF-kappa B activity. I kappa B is phosphorylated by I kappa B kinase (IKK) complex that contains IKK alpha , IKK beta , and IKK gamma. A novel molecule that associates with and activates IKK was recently identified and designated CIKS (for connection to IKK and SAPK/JNK) and Act1 (for NF-kappa B activator 1). CIKS directly interacts with IKK gamma. CIKS/Act1 also activates activating transcription factor (ATF) and activator protein 1 (AP-1) through Jun kinase (JNK). These results indicate that CIKS/Act1 is involved in the inflammation and stress responses. CIKS/Act1 is ubiquitously expressed in human tissues.
Type:
Antigen: CIKS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: