- Antigen name:G Protein-coupled Receptor 15
- Antigen symbol:GPR15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:GPR15|BOB
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:GPR15 antibody was raised against a peptide corresponding to amino acids near the amino terminus of human GPR15. The sequence differs from those of African green monkey and pig-tailed macaque BOB by one amino acid (2).
- Tested applications:E, WB
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
GPR15 Antibody: Human immunodeficiency virus (HIV) and related viruses require coreceptors to infect target cells. Some G protein-coupled receptors including CCR5, CXCR4, CCR3, CCR2b and CCR8 in the chemokine receptor family, and four new human molecules GPR15, STRL33, GPR1 and V28 were recently identified as HIV coreceptors. The genes encoding human and monkey GPR15/BOB (for G protein-coupled receptor 15 and brother of Bonzo, respectively, ) were recently cloned. This novel G protein-coupled receptor serves as corecptor for simian immunodeficiency virus (SIV), and for strains of HIV-2 and M-tropic HIV-1. The ligand for GPR15 has not been identified yet.
Type:
Antigen: GPR15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat