TE buffer, 1X, ready to use, pH 8,0, low EDTA, Molecular Biology Grade, Affynmetrix/USB
Supplier: Thermo Fisher Scientific
Certificates
About this item
TE buffer with low EDTA. Use directly to re-suspend and/or dilute purified DNA or RNA.
- 1X TE solution consisting of 10 mM Tris Ultrapure, pH 8,0 and 0,1 mM EDTA
- Solution is 0,2 μm filtered and dispensed into vials and/or durable, square Nalgene bottles
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together