VWR®, Industrial Wipes, Clean Smooth, White
Supplier: VWR Collection
Certificates
About this item
These bleached, spunlace wipes, made from 55% wood pulp and 45% polyester, are extremely strong, even when wet.
- Low fibre and particle release
- Resistant to acids, solvents and chlorine
- Good absorption
- Do not contain any binders, silicone-free
- Recyclable
- Food contact certified
Typical applications include: Pharma, aerospace, semiconductor, food and beverage, automotive, laboratories, printing and healthcare.
Basis weight: 68 g/m²
Absorption: 400%