VWR®, Deep Well Plates, 96 Square Well, Reinforced for Genomics Applications
Supplier: VWR Collection
Certificates
About this item
With reinforced walls to the bottom of the well backed, and supporting ribs on the underside of the plate, these 2 ml square well PP blocks can be used safely and with confidence for plant DNA extraction. The wells have a total volume of 2,12 ml giving a generous working volume of 1,90 ml per well in a 45 mm high plate.
- Made from pure, virgin, extractable-free PP, rigorously tested for leachates
- Solid, stable and strong – able to withstand the applied forces in bead beaters
- Manufactured to ANSI/SLAS specifications
- Free from DNase and RNase
Accessories information: Re-usable friction seals are available for these plates. The standard EVA cap mat is flexible and easy to apply. For those wishing to freeze the sealed plate at –80 °C, a special PP cap mat is available. This forms a very tight seal, and because it is made of the same polymer as the plate, it expands and shrinks at the same rate, thereby ensuring that the cap mat will not loosen as the temperature changes, preventing cross-contamination between wells. A special cap mat applicator is also available designed to reduce the risk of repetitive strain injuries whilst sealing large numbers of plates.