Poly(diallyldimethylammonium chloride) [pDADMAC] 10% in water flocculation reagent, Millipore®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
Synonyms: Polyquaternium-6, N,N-Dimethyl-N-2-propenyl-2-propen-1-aminiumchloridehomopolymer, PolyDADMAC
CAS Number:26062-79-3
:
Formula:(C₈H₁₆ClN)n
MDL Number:MFCD00192409
Density:1.015 g/cm3
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity (NIR):Passes test
- Appearance:Colourless to yelowish liquid
- Dry residue (105 °C):9.0 - 11.0%
- Total aerobic microbial count (TAMC):≤10 CFU
- Total yeast and mould count (TYMC):≤10 CFU
- Bile tolerant gram negative bacteria:absent in 1 ml
- candida albicans:absent in 1 ml
- E. coli:absent in 1 ml
- Pseudomonas aeruginosa:absent in 1 ml
- Salmonella species:absent in 10 ml
- Staphylococcus aureus:absent in 1 ml
- Endotoxins:≤500 I.U/ml
- Monomer content:≤0.1%
- Viscosity (20 °C):100 - 350 mPa.s
Specification Test Results
Frequently Bought Together