Order Entry
Denmark
ContactUsLinkComponent
Anti-SENP5 Rabbit Polyclonal Antibody (Cy5.5®)
Anti-SENP5 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-8464R-CY5.5
Supplier:  Bioss
Anti-SENP5 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-8464R-CY5.5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sentrin-specific Protease 5
  • Antigen symbol:
    SENP5
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    205564
  • Antigen synonyms:
    Sentrin/SUMO-specific protease SENP5|Sentrin-specific protease 5|FKSG45
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Protease that catalyzes two essential functions in the SUMO pathway: processing of full-length SUMO3 to its mature form and deconjugation of SUMO2 and SUMO3 from targeted proteins. Has weak proteolytic activity against full-length SUMO1 or SUMO1 conjugates. Required for cell division.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SENP5
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat