Anti-GK5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-8408R-A555
: Bioss
- Antibody type:Primary
- Antigen name:Glycerol Kinase 5
- Antigen symbol:GK5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:256356
- Antigen synonyms:Putative glycerol kinase 5.|FLJ33582|ATP:glycerol 3-phosphotransferase 5|glycerol kinase 5 (putative)|MGC40579|Glycerokinase 5|FLJ45739
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
GK5
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GK5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit