Order Entry
Denmark
ContactUsLinkComponent
Anti-COLEC10 Rabbit Polyclonal Antibody (Cy5®)
Anti-COLEC10 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-8033R-CY5
Supplier:  Bioss
Anti-COLEC10 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-8033R-CY5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Collectin sub-family member 10
  • Antigen symbol:
    COLEC10
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    10584
  • Antigen synonyms:
    CLL1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    142-187/277
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Lectin that binds to various sugars: galactose >mannose = fucose >N-acetylglucosamine >N-acetylgalactosamine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COLEC10
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity: