Anti-A4GALT Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-7588R-HRP
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Alpha 1,4-Galactosyltransferase
- Antigen symbol:A4GALT
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:53947
- Antigen synonyms:A4GALT1|P(k)|P1|P1PK|Gb3S|A4GALT|PK|A14GALT
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:290-340/353
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Necessary for the biosynthesis of the Pk antigen of blood histogroup P. Catalyzes the transfer of galactose to lactosylceramide and galactosylceramide. Necessary for the synthesis of the receptor for bacterial verotoxins.
Recommended Dilutions: IHC-P: 1:100-500
Type: Primary
Antigen: A4GALT
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat