Anti-GAS6 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-7549R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Growth Arrest Specific 6
- Antigen symbol:GAS6
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:2621
- Antigen synonyms:AXSF|Gas-6|AXLLG
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:560-610/721
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Ligand for tyrosine-protein kinase receptors AXL, TYRO3 and MER whose signaling is implicated in cell growth and survival, cell adhesion and cell migration. GAS6/AXL signaling plays a role in various processes such as endothelial cell survival during acidification by preventing apoptosis, optimal cytokine signaling during human natural killer cell development, hepatic regeneration, gonadotropin-releasing hormone neuron survival and migration, platelet activation, or regulation of thrombotic responses.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GAS 6
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat