Anti-BTG1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-7526R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:B-cell translocation gene 1, anti-proliferative
- Antigen symbol:BTG1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:694
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
BTG1 belongs to the BTG family of proteins.FUNCTION: Anti-proliferative protein. Its expression is associated with the early G1 phase of the cell cycle. A chromosomal aberration involving BTG1 may be a cause of a form of B cell chronic lymphocytic leukemia; translocation t(8;12)(q24;q22) with MYC.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BTG1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat