Anti-DUSP22 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-7066R-HRP
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Dual Specificity Phosphatase 22
- Antigen symbol:DUSP22
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:56940
- Antigen synonyms:JSP1|LMWDSP2|MKPX|LMW-DSP2|JKAP|MKP-x|JSP-1|VHX
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Activates the Jnk signaling pathway. Dephosphorylates and deactivates p38 and stress-activated protein kinase /c-Jun N-terminal kinase (SAPK/JNK)
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: DUSP22
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat