Specifications
- Antibody type:Primary
- Antigen name:Nuclear receptor-interacting protein 1
- Antigen symbol:NRIP
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:8204
- Antigen synonyms:DCAF6_HUMAN.|IQWD1v MSTP055|NRIP1|ARCAP|IQ mot and WD repeats 1|Androgen receptor complex associated protein|Nuclear receptor interaction protein|RP4 745I14.1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
WD-repeat proteins are a large family of eukaryotic proteins coordinating multi-protein complex assemblies. Their role has been implicated in multiple cellular processes including signal transduction, transcriptional regulation, cell cycle control and apoptosis. NRIP is a novel 860a.a nuclear protein consisting of seven conserved WD40 domains and one NLS motif. It binds to androgen and glucocorticoid receptors and up-regulates their transcriptional activity, thereby functioning as a nuclear receptor co-activator. Role of NRIP has been implicated in cell growth and also in cervical and prostrate cancer, thus indicating a potential therapeutic intervention. Northern Blot analysis detects a high expression of NRIP in skeletal muscle and testis and low expression in heart, prostrate and adrenal gland.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NRIP
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat