Anti-FBXO31 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-6006R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:F-Box Protein 31
- Antigen symbol:FBXO31
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:79791
- Antigen synonyms:MRT45|Fbx31|FBXO14|FBX14|pp2386
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
FBXO31 belongs to the F-box protein family. Such proteins are characterized by an F-box motif of approximately 40 residues. F-box proteins interact with SKP1 through the F box and they interact with ubiquitination targets through other protein interaction domains. There are two different isoforms.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FBXO31
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat