Anti-MAP7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-5718R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Microtubule-Associated Protein 7
- Antigen symbol:MAP7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:9053
- Antigen synonyms:EMAP115|E-MAP-115
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:160-210/749
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Microtubule-stabilizing protein that may play an important role during reorganization of microtubules during polarization and differentiation of epithelial cells. Associates with microtubules in a dynamic manner. May play a role in the formation of intercellular contacts. Colocalization with TRPV4 results in the redistribution of TRPV4 toward the membrane and may link cytoskeletal microfilaments.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MAP7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat