Order Entry
Denmark
ContactUsLinkComponent
Anti-CDC25A Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-CDC25A Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-5242R-A350
Supplier:  Bioss
Anti-CDC25A Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-5242R-A350
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cell Division Cycle 25A
  • Antigen symbol:
    CDC25A
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    993
  • Antigen synonyms:
    Dual specificity phosphatase Cdc25A|CDC25A|M-phase inducer phosphatase 1
  • Modification:
    phospho-Ser124
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Cyclin dependent kinases are positively regulated by association with cyclins and negatively regulated by binding to inhibitory subunits. The activity of cyclin dependent kinases is also regulated by the phosphorylation status, which is controlled by the antagonistic action of Wee1 kinase and CDC25 phosphatases. Three CDC25 genes are present in human cells: CDC25A, CDC25B, and CDC25C. These three genes function at different phases of the cell cycle. Whereas CDC25A and CDC25B are expressed throughout the cell cycle, with peak expression in G1 for CDC25A and in both G1 S phase and G2 for CDC25B, CDC25C is predominantly expressed in G2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDC25A
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: