Specifications
- Antibody type:Primary
- Antigen name:Prion Protein
- Antigen symbol:PRNP
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:PRIP|Sinc.|PrP33 35C|CD230 antigen|ASCR|PrP27 30|KURU|AltPrP|p27 30|Creutzfeld Jakob disease|Major prion protein|CD230|PrP27-30|PrP33-35C|GSS|Prnp|atal familial insomnia|Gerstmann-Strausler-Scheinker syndrome|PRIO_HUMAN|PrPSc|MGC26679|PrPC|PrP|Prni|Prion related protein|CJD|Prion protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The protein encoded by this gene is a membrane glycosylphosphatidylinositol-anchored glycoprotein that tends to aggregate into rod-like structures. The encoded protein contains a highly unstable region of five tandem octapeptide repeats. This gene is found on chromosome 20, approximately 20 kbp upstream of a gene which encodes a biochemically and structurally similar protein to the one encoded by this gene. Mutations in the repeat region as well as elsewhere in this gene have been associated with Creutzfeldt-Jakob disease, fatal familial insomnia, Gerstmann-Straussler disease, Huntington disease-like 1, and kuru. An overlapping open reading frame has been found for this gene that encodes a smaller, structurally unrelated protein, AltPrp. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Oct 2012].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PRNP
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat