Order Entry
Denmark
ContactUsLinkComponent
 
Anti-PNPLA2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-3831R-A488
 :  Bioss
Anti-PNPLA2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-3831R-A488
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Patatin-like Phospholipase Domain Containing 2
  • Antigen symbol:
    PNPLA2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    57104
  • Antigen synonyms:
    iPLA2zeta|TTS-2.2|ATGL|TTS2|FP17548|PEDF-R|Desnutrin
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    415-445/504
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Catalyzes the initial step in triglyceride hydrolysis in adipocyte and non-adipocyte lipid droplets. Also has acylglycerol transacylase activity. May act coordinately with LIPE/HLS within the lipolytic cascade. Regulates adiposome size and may be involved in the degradation of adiposomes. May play an important role in energy homeostasis. May play a role in the response of the organism to starvation, enhancing hydrolysis of triglycerides and providing free fatty acids to other tissues to be oxidized in situations of energy depletion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PNPLA2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat