Anti-HMGN1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-4113R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:High-mobility Group Nucleosome Binding Domain 1
- Antigen symbol:HMGN1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:3150
- Antigen synonyms:MGC117425|FLJ31471|MGC104230|HMG14|FLJ27265
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The protein encoded by this gene binds nucleosomal DNA and is associated with transcriptionally active chromatin. Along with a similar protein, HMG17, the encoded protein may help maintain an open chromatin configuration around transcribable genes. [provided by RefSeq, Aug 2011]
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HMGN1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat