Anti-ATG4A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-3873R-A647
: Bioss
- Antistof type:Primary
- Antigen name:Autophagy Related 4A
- Antigen symbol:ATG4A
- Clonality:Polyclonal
- Konjugation:Alexa Fluor® 647
- Vært:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Krydsadsorption:No
- Form:liquid
- Gen ID:115201
- Antigen synonymer:APG4A|autophagin-2|Cysteine protease ATG4A|AUTL2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Opbevaringstemperatur:Store at 4°C for 12 months
- Koncentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pakn:100 µl
ATG4A is a cysteine protease required for autophagy, which cleaves the C-terminal part of either MAP1LC3, GABARAPL2 or GABARAP, allowing the liberation of form I. A subpopulation of form I is subsequently converted to a smaller form (form II). Form II, with a revealed C-terminal glycine, is considered to be the phosphatidylethanolamine (PE)-conjugated form, and has the capacity for the binding to autophagosomes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ATG4A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat