Anti-HSF1 Rabbit Polyclonal Antibody (Cy7®)
BOSSBS-3757R-CY7
: Bioss
- Antibody type:Primary
- Antigen name:Heat Shock Transcription Factor 1
- Antigen symbol:HSF1
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:3297
- Antigen synonyms:HSTF 1|Heat shock transcription factor 1|Heat shock factor protein 1|HSTF1|HSF 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:360-410/529
- Purification:Purified by Protein A
- Pk:100 µl
DNA-binding protein that specifically binds heat shock promoter elements (HSE) and activates transcription. In higher eukaryotes, HSF is unable to bind to the HSE unless the cells are heat shocked.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HSF1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat