Anti-CYBA Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-3879R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome B245 alpha
- Antigen symbol:CYBA
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:1535
- Antigen synonyms:p22-PHOX|Cytochrome b(558) alpha chain
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:30-80/195
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Critical component of the membrane-bound oxidase of phagocytes that generates superoxide. Associates with NOX3 to form a functional NADPH oxidase constitutively generating superoxide.
Recommended Dilutions: IF(IHC-P): 1:50-200; IF(IHC-F)(1:50-200
Type: Primary
Antigen: CYBA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity: