Anti-GAPDHS Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-13282R-A647
: Bioss
- Antibody type:Primary
- Antigen name:Sperm-specific glyceraldehyde phosphate dehydrogenase
- Antigen symbol:GAPDHS
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:26330
- Antigen synonyms:GAPDH-2|HSD-35|GAPD2|GAPDS|GAPDH S
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:275-325/408
- Purification:Purified by Protein A
- Pk:100 µl
May play an important role in regulating the switch between different pathways for energy production during spermiogenesis and in the spermatozoon. Required for sperm motility and male fertility (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GAPDHS/GAPDH2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat