Specifikationer
- Antistof type:Primary
- Antigen name:Cadherin 1
- Antigen symbol:CDH1
- Clonality:Polyclonal
- Konjugation:Cy5®
- Vært:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Krydsadsorption:No
- Form:liquid
- Gen ID:999
- Antigen synonymer:CD324|epithelial Cadherin|Cadherin-E|ECAD|Arc-1|CAM 120/80|cell-CAM 120/80|E-Cadherin|uvomorulin|Cadherin 1|Cadherin-1|E-Cadherin epithelial|calcium-dependent adhesion protein|epithelial|LCAM|CDHE|UVO
- Modification:phospho-Ser838/Ser840
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Opbevaringstemperatur:Store at 4°C for 12 months
- Koncentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pakn:100 µl
Specifikationer
Om denne vare
Cadherins comprise a family of Ca2+-dependent adhesion molecules that function to mediate cell-cell binding critical to the maintenance of tissue structure and morphogenesis. Members of this family of adhesion proteins include rat cadherin K (and its human homolog, cadherin-6), R-cadherin, B-cadherin, E/P cadherin and cadherin-5. The classical cadherins, E-, N- and P-cadherin, consist of large extracellular domains characterized by a series of five homologous NH2 terminal repeats. The most distal of these cadherins is thought to be responsible for binding specificity, transmembrane domains and carboxy terminal intracellular domains. The relatively short intracellular domains interact with a variety of cytoplasmic proteins, such as ∫-catenin, to regulate cadherin function.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CDH1
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: