Specifications
- Antibody type:Primary
- Antigen name:Acyl-CoA Binding Domain Containing 3
- Antigen symbol:ACBD3
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:GOCAP 1|Peripherial benzodiazepine receptor associated protein|PAP 7|ACBD 3|Golgi complex associated protein 1 60kDa|GCP60|Golgi phosphoprotein 1|GCP 60|Golgi complex-associated protein 1|Acyl Coenzyme A binding domain containing 3|GOLPH 1|GOCAP1|Acyl CoA binding domain containing protein 3|Peripheral benzodiazepine receptor associated protein PAP7|PKA RIalpha associated protein.|PBR associated protein|ACBD3|PAP7|Golgi complex associated protein 1|GCP60_HUMAN|PBR and PKA associated protein 7|Golgi resident protein GCP60|PBR- and PKA-associated protein 7|Acyl-CoA-binding domain-containing protein 3|Peripheral benzodiazepine receptor-associated protein PAP7|GOLPH1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
ACBD3 (acyl-CoA-binding domain-containing protein 3), also known as GCP60 (Golgi resident protein GCP60), GOCAP1, PAP7 or GOLPH1, is a Golgi apparatus membrane protein that contains one ACB (acyl-CoA-binding) domain and one GOLD (Golgi dynamics) domain which is essential for its interaction with other proteins. Expressed ubiquitously with highest expression in ovary and testis, ACBD3 is responsible for maintaining Golgi structure and, through binding to Giantin (golgin subfamily B member 1), functions to mediate protein transport between the Golgi and the endoplasmic reticulum (ER). Changes in the subcellular location of ACBD3 trigger signaling cascades within the Golgi that regulate cell fate and cell cycle progression. Additionally, ACBD3 is thought to act as a peripheral-type benzodiazepine receptor-associated protein, possibly playing a role in hormonal regulation and steroid formation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ACBD3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat