Anti-CUX2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-11832R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Cut-like homeobox 2
- Antigen symbol:CUX2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:23316
- Antigen synonyms:CUTL2|CDP2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:1030-1080/1486
- Purification:Purified by Protein A
- Pk:100 µl
May be a transcription factor involved in neural specification. Binds to DNA in a sequence-specific manner (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CUX2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: