Specifications
- Antibody type:Primary
- Antigen name:Germ Cell Nuclear Factor
- Antigen symbol:GCNF
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Retinoid receptor related testis specic receptor|RTR.|Orphan nuclear receptor NR6A1|NR6A1_HUMAN|NR61|NR 61|NR6A1|Germ cell nuclear factor|GCNF|Nuclear receptor subfamily 6 group A member 1|GCNF 1|hRTR|hGCNF|Retinoic acid receptor related testis associated receptor|Retinoid receptor-related testis-specic receptor|GCNF1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Germ cell nuclear factor (GCNF) is an orphan member of the nuclear receptor gene superfamily that influences neurogenesis and germ cell development. GCNF can homodimerize and bind DNA. GCNF regulates paracrine interaction between the oocyte and somatic cells by regulating the expression of BMP-15 and GDF-9, to affect female fertility. GCNF is present in spermatocytes and round spermatids of adult male mouse testis; northern blot and ribonuclease protection assays have shown GCNF is predominant in the testis. The gene expresses three alternatively spliced transcript variants.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GCNF
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat