Specifications
- Antibody type:Primary
- Antigen name:Synaptojanin 1
- Antigen symbol:SYNJ1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:8867
- Antigen synonyms:inositol 5'-phosphatase (synaptojanin 1)|synaptojanin-1|synaptic inositol-1,4,5-trisphosphate 5-phosphatase 1|polyphosphoinositide phosphatase|INPP5G
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The inositol polyphosphate 5-phosphatases selectively remove the phosphate from the 5-position of various phosphatidylinositols, which generate second messengers in response to extracellular signals. Synaptojanins are characterized by an N-terminal SAC1-like sequence, a central 5-phosphate domain, and a unique C-terminal sequence and have been shown to use phosphatidylinositol 4,5-bisphosphate as a substrate. Synaptojanins exist as two isoforms, synaptojanin 1 and 2, which differ in the C-terminal domain, and each isoform has multiple variants produced by alternative splicing. Synaptojanin 1 is expressed as two major forms: the shorter is found in brain while the longer is expressed in peripheral tissues. Eight splice variants of synaptojanin 2 have been detected, including a brain specific isoform. Synaptojanins are thought to participate in the endocytosis of synaptic vesicles and the regulation of the actin cytoskeleton.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SYNJ1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat